Learn More
Medchemexpress LLC PDGFR Tyrosine Kinase Inhibitor III | 205254-94-0 | >99.0% | 485.53 | 10 MG
Supplier: Medchemexpress LLC HY11241210MG
PDGFR Tyrosine Kinase Inhibitor III is a multikinase inhibitor that targets and inhibits multiple receptor tyrosine kinases. It specifically inhibits Platelet-Derived Growth Factor Receptor (PDGFR), Epidermal Growth Factor Receptor (EGFR), Fibroblast Growth Factor Receptor (FGFR), Protein Kinase A (PKA), and Protein Kinase C (PKC). This compound demonstrates potential in various research applications, particularly in studies related to amyotrophic lateral sclerosis.
- Acts as a multikinase inhibitor.
- Inhibits PDGFR, EGFR, FGFR, PKA, and PKC.
- Suitable for research into amyotrophic lateral sclerosis.