Learn More
ABclonal Technology PDHA2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A149941000UL
Encompass_Preferred
Predicted to enable pyruvate dehydrogenase (acetyl-transferring) activity. Predicted to contribute to pyruvate dehydrogenase (NAD+) activity. Predicted to be involved in acetyl-CoA biosynthetic process from pyruvate. Predicted to act upstream of or within mitochondrial acetyl-CoA biosynthetic process from pyruvate. Located in mitochondrion. Is expressed in nervous system and testis. Orthologous to human PDHA2 (pyruvate dehydrogenase E1 subunit alpha 2).
Catalog No. 50-001-06841