Learn More
Sino Biological Peptide Substrates: PDHKtide
Supplier:  Sino Biological P5758
Encompass_Preferred
The synthetic peptide substrate PDHKtide (RRYHGHSMS-DPGVSYRTR) is derived from human PDHA1 protein (amino acid 288-304aa) and can be used for PDHK family kinase assay.
Catalog No. 50-256-8164