Learn More
BEZWADA BIOMEDICAL LLC Poly(D,L-lactide-co-glycolide), D,L-lactide:glycolide 85:15, viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 4281
Encompass_Preferred
In order to decrease the absorption times of implantable devices incorporating L(-)-lactide, copolymers of L(-)-lactide and dl-lactide with glycolide have been prepared. The polymers are used in medical devices and drug delivery.
Catalog No. 50-190-4475