Learn More
Cayman Chemical PDM 11 Fatty Acids and Derivat
SDP
Supplier:  Cayman Chemical 1000634125 MG
Encompass_Preferred
A derivative of resveratrol; inactive in vitro in assays of resveratrol activity; does not affect proliferation of K562, HT-29, and HepG2 cells
Catalog No. 50-288-8161