Learn More
Medchemexpress LLC PDP-Pfp | 160580-70-1 | 98.3% | 381.34 g/mol | C14H8F5NO2S2 | 500 MG

Supplier: Medchemexpress LLC HY129359500MG
PDP-Pfp is a reducible antibody-drug conjugate (ADC) linker intended for use with agents that target the extracellular loop 1 (ECL1) of TM4SF1. It is supplied as an off-white powder with defined molecular properties and analytical purity suitable for ADC research and development.
- Reducible linker chemistry suitable for disulfide-based ADCs.
- High chemical purity (98.3%).
- Stable as powder at -20°C for long-term storage.
- Solvent solutions: store at -80°C for up to 6 months or -20°C for up to 1 month.
- Available in multiple pack sizes, including 500 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.