Learn More
Medchemexpress LLC Pds-MMAE (Compound 5') | 1613113-44-2 | 99.8% | 931.26 | C47H74N6O9S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY15395825MG
Encompass_Preferred
PDS-MMAE (Compound 5') is a modified monomethyl auristatin E (MMAE) supplied as a research reagent for preparation of receptor-binding conjugates, including neurotensin receptor binding conjugates. It is produced at high purity and offered in milligram-scale quantities for conjugation and biochemical research.
  • High purity (99.8%).
  • Suitable for conjugation to peptides and proteins.
  • Available in multiple milligram-scale pack sizes.
  • Store at 4°C and protect from light; stable in solvent at -80°C for up to 6 months.
Catalog No. 50-002-04354