Learn More
Biolegend PE anti-β Catenin 1
Supplier:  Biolegend 862604
Encompass_Preferred
PE anti-β Catenin 1 [15B8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: ICFC; Size: 100 tests
Catalog No. 50-207-4892