Learn More
Biolegend PE anti-human CD152 (CTLA-4)
Supplier:  Biolegend 349905
Encompass_Preferred
PE anti-human CD152 (CTLA-4) [L3D10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-168-555