Learn More
Biolegend PE anti-human CD272 (BTLA)
Supplier:  Biolegend 344505
Encompass_Preferred
PE anti-human CD272 (BTLA) [MIH26]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green; Apps: FC; Size: 25 tests
Catalog No. 50-168-250