Learn More
Biolegend PE anti-human CD294 (CRTH2)
Supplier:  Biolegend 350106
Encompass_Preferred
PE anti-human CD294 (CRTH2) [BM16]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-576