Learn More
Biolegend PE anti-human CD317 (BST2, Tetherin)
Supplier: Biolegend 348405
PE anti-human CD317 (BST2, Tetherin) [RS38E]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Pigtail, Rhesus Macaque; Apps: FC; Size: 25 tests