Learn More
Biolegend PE anti-human CD34
Supplier:  Biolegend 343606
Encompass_Preferred
PE anti-human CD34 [561]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-209