Learn More
Biolegend PE anti-human CD74
Supplier:  Biolegend 326807
Encompass_Preferred
PE anti-human CD74 [LN2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-343