Learn More
Biolegend PE anti-human CD79b (Igβ)
Supplier:  Biolegend 341404
Encompass_Preferred
PE anti-human CD79b (Igβ) [CB3-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-069