Learn More
Biolegend PE anti-human Delta-like protein 4 (DLL4)
Supplier:  Biolegend 346506
Encompass_Preferred
PE anti-human Delta-like protein 4 (DLL4) [MHD4-46]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-380