Learn More
Biolegend PE anti-human Galectin-9
Supplier: Biolegend 348905
PE anti-human Galectin-9 [9M1-3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: ICFC, FC; Size: 25 tests