Learn More
Biolegend PE anti-human HLA-A,B,C
Supplier:  Biolegend 311406
Encompass_Preferred
PE anti-human HLA-A,B,C [W6/32]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-629