Learn More
Biolegend PE anti-human TCR Vα7.2
Supplier:  Biolegend 351705
Encompass_Preferred
PE anti-human TCR Vα7.2 [3C10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-168-676