Learn More
Biolegend PE anti-human TLT-2
Supplier:  Biolegend 351104
Encompass_Preferred
PE anti-human TLT-2 [MIH61]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-637