Learn More
Biolegend PE anti-human TLT-2
Supplier:  Biolegend 351204
Encompass_Preferred
PE anti-human TLT-2 [MIH60]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-641