Learn More
Biolegend PE anti-human CD65
Supplier:  Biolegend 351404
Encompass_Preferred
PE anti-human CD65 [VIM8]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-286-5986