Learn More
Biolegend PE anti-human HLA-G Recombinan
Supplier:  Biolegend 381804
Encompass_Preferred
PE anti-human HLA-G Recombinant [QA19A11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2034