Learn More
Biolegend PE anti-human LGR5 (GPR49)
Supplier:  Biolegend 373804
Encompass_Preferred
PE anti-human LGR5 (GPR49) [SA222C5]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8778