Learn More
Biolegend PE anti-mouse CD369 Dectin-1 C
Supplier:  Biolegend 144303
Encompass_Preferred
PE anti-mouse CD369 (Dectin-1, CLEC7A) [RH1]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2659