Learn More
Biolegend PE anti-mouse Podoplanin [8.1.1]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Supplier:  Biolegend 127407
Encompass_Preferred
PE anti-mouse Podoplanin [8.1.1]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-368