Learn More
Biolegend PE anti-mouse CD3ε
Supplier:  Biolegend 100308
Encompass_Preferred
PE anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-162-388