Learn More
Biolegend PE anti-mouse CD41
Supplier:  Biolegend 133905
Encompass_Preferred
PE anti-mouse CD41 [MWReg30]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-704