Learn More
Biolegend PE anti-mouse FOXP3
Supplier:  Biolegend 126404
Encompass_Preferred
PE anti-mouse FOXP3 [MF-14]; Isotype: Rat IgG2b, κ; Reactivity: mouse; Apps: ICFC; Size: 100 ug
Catalog No. 50-164-291