Learn More
Biolegend PE anti-mouse H-2Kb/H-2Db
Supplier:  Biolegend 114607
Encompass_Preferred
PE anti-mouse H-2Kb/H-2Db [28-8-6]; Isotype: Mouse (C3H) IgG2a, κ; Reactivity: Mouse b haplotype, Cross-reacts with H-2Dd MHC class I alloantigen; Apps: FC; Size: 50 ug
Catalog No. 50-163-501