Learn More
Biolegend PE anti-mouse H-2Kk
Supplier:  Biolegend 114907
Encompass_Preferred
PE anti-mouse H-2Kk [36-7-5]; Isotype: Mouse (A.TL) IgG2a, κ; Reactivity: Mouse k haplotype, Weakly cross-reacts with SJL/Hsd mice splenocytes; Apps: FC; Size: 50 ug
Catalog No. 50-163-516