Learn More
Biolegend PE anti-mouse lymphotoxin beta receptor (LTβR)
Supplier:  Biolegend 134403
Encompass_Preferred
PE anti-mouse lymphotoxin beta receptor (LTβR) [5G11]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-746