Learn More
Biolegend PE anti-mouse TCR DO11.10
Supplier:  Biolegend 118508
Encompass_Preferred
PE anti-mouse TCR DO11.10 [KJ1-26]; Isotype: Mouse IgG2a, κ; Reactivity: mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-786