Learn More
Biolegend PE/Cyanine7 anti-human CD138 (Syndecan-1)
Supplier:  Biolegend 356513
Encompass_Preferred
PE/Cyanine7 anti-human CD138 (Syndecan-1) [MI15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7543