Learn More
Alpco Ltd Pancreatic Elastase Chemiluminescence ELISA
SDP
Supplier:  Alpco Ltd 80PANHUCH01
Encompass_Preferred
The ALPCO Pancreatic Elastase (PE) Chemiluminescence ELISA is a chemiluminescent immunoassay for the quantitative determination of pancreatic elastase in human stool. Pancreatic elastase (PE or PE-1) refers to a family of chymotrypsin-like elastases (CELAs), digestive proteases produced by acinar cells in the pancreas. PE is highly stable in the intestinal tract and the concentration of pancreatic elastase in stool correlates well with overall pancreatic enzyme output. Both the CELA3A and CELA3A isoforms of PE can be detected in stool and are detected by the Pancreatic Elastase Chemiluminescence ELISA from ALPCO. FDA Registered. For In Vitro Diagnostic Use. Key features: User-friendly workflow featuring ready-to-use standards and controls, Broad analytical measuring range, Standardized to AAA-tested, stool-derived PE. Compatible with the 30-EZEX-100 stool extraction devices and 10-EXBUF-55 (or 10-EXBUF-225) stool extraction buffer.
Catalog No. NC2271702