Learn More
MSC PIP CoreShield 3.0 Cut-Resistant Gloves, A4 Cut, 15 Gauge, X-Large, 1 Pair
Supplier:  MSC 42100255
Encompass
These 15-gauge cut-resistant gloves offer reliable protection for heavy-duty tasks in the automotive, manufacturing, and metalworking industries. Constructed with a fiberglass-free liner from recycled polyester and carbon fiber, they provide ANSI/ISEA A4 cut resistance and feature a durable, breathable nitrile micro-foam palm coating for superior grip in dry, wet, and oily conditions.
  • Provides reliable dexterity with a 15-gauge fiberglass-free liner
  • Features a breathable nitrile micro-foam coating to disperse fluids for improved grip
  • Includes a reinforced thumb crotch to enhance durability in high-wear areas
  • Complies with FDA 21 CFR, Part 177 for food handling
  • ESD-safe construction allows for use with static-sensitive materials
  • Touchscreen compatible for device operation without glove removal
  • Oeko-Tex® standard 100 certified for safety against skin irritation
Catalog No. 50-005-08854