Learn More
MSC Honeywell Cut Resistant Gloves, 15 gg, Nitrile Coated, Size 9, 10 PR
Supplier:  MSC 37102126
Encompass
These CoreShield 3.0 cut resistant gloves provide advanced hand protection for diverse industrial environments. Designed with a 15 gauge seamless liner, they offer a precise fit and dexterity for handling sharp objects while maintaining tactile sensitivity.
  • Features 15 gauge seamlessly knitted recycled polyester, polyethylene and carbon fibre liner
  • Utilizes nitrile micro-foam coating for high grip performance and breathability
  • Provides cut level C/A3 protection according to ANSI/ISEA 105 and EN 388:2016 standards
  • Includes reinforced thumb for increased durability in high-wear areas
  • Antistatic and silicone-free construction with low DMF content
  • Certified for food handling and touch screen operation
  • Sustainable design using recycled fibres and plastic-free paper packaging
  • Washable for extended utility
Catalog No. 50-005-08835