Learn More
MSC PIP G-Tek® CoreShield® Seamless Knit Gloves, HPPE, Cut Level A2, Size 11, 100/Cs.
Supplier: MSC 37096682
These seamless knit engineered yarn gloves are designed for comfort and reliable hand protection in environments requiring cut resistance. Featuring a foam nitrile coating, they offer enhanced grip and durability for various handling tasks.
- Provides A2 cut level protection
- Features touchscreen compatibility for device interaction
- Designed with a reinforced thumb crotch for extended wear
- Includes ESD safe properties for work in sensitive areas
- Constructed with a knit wrist for a secure fit
- Coating covers palm, fingers and knuckles
- Meets EN 388 2016 standard with 4X42B results