Learn More
MSC PIP CoreShield 3.0 Cut-Resistant Gloves, 15 Gauge, Size 6, 1/Pr.
Supplier: MSC 37101821
These 15 gauge gloves are designed for comfort and high-level protection in industrial applications. Featuring a reinforced thumb and seamless construction, they offer a comfortable fit while maintaining durability. The gloves are treated with a specialized nitrile foam coating on the palm for enhanced grip in various working conditions and comply with multiple safety standards.
- Nitrile micro-foam palm coating for superior grip
- Cut level C/A3 for hand protection
- Fiberglass-free construction for safety
- Reinforced thumb for increased durability
- Seamlessly knitted design for reduced hand fatigue
- Suitable for automotive, assembly, and maintenance applications
- Compliant with EN388 4X42C and EN 16350 standards
- Approved for food contact per FDA 21CFR177.2600 and EC 1935/2004