Learn More
MSC PIP Cut Resistant Gloves, 15 Gauge, HPPE, Nitrile Coated, XL, 100/Case
Supplier:  MSC 37102407
Encompass
These cut-resistant gloves feature a 15-gauge, fiberglass-free HPPE liner designed for reliable dexterity, comfort, and breathability. They offer an ANSI A3 cut resistance level and are coated with foam nitrile for an improved grip in various applications, while the ESD-safe, silicone-free construction makes them suitable for use with static-sensitive materials and smudge-free handling.
  • Seamless construction provides increased comfort
  • Foam nitrile coating disperses fluids for improved grip
  • Touchscreen compatible design allows device operation without removal
  • Reinforced thumb crotch enhances durability in high-wear areas
  • Complies with FDA 21 CFR, Part 177 food handling requirements
  • Oeko-Tex® standard 100 certified for skin safety
  • Vend-ready packaging for efficient dispensing
Catalog No. 50-005-08668