Learn More
MSC Honeywell CoreShield 3.0 Cut Resistant Gloves, Nitrile, A3, 15 gg, 1/Pr.
Supplier: MSC 37101763
These 15-gauge cut-resistant gloves provide advanced hand protection through a lightweight, ergonomically shaped design. Featuring a seamless liner made from recycled polyester, polyethylene (PE), carbon fiber, and spandex, these gloves are designed for comfort and dexterity in demanding applications. The nitrile micro-foam coating provides exceptional grip in both wet and dry conditions, while the integrated thumb reinforcement enhances durability.
- Cut resistance level A3 (ANSI) and C (EN 388)
- Glass fiber-free construction
- Certified for food handling under FDA 21CFR177.2600 and EC regulations
- Antistatic properties tested to EN 16350
- Touchscreen compatible for ease of use
- Silicone-free and low DMF content
- Washable for extended service life
- Color-coded for efficient sizing identification