Learn More
MSC Pip CoreShield 3.0 Cut Resistant Gloves, HPPE, Nitrile, A3, 1 Pair
Supplier:  MSC 37103033
Encompass
These CoreShield™ 3.0 cut protection gloves are designed for high-performance safety in environments such as automotive and parts manufacturing. Featuring an 18-gauge seamless knit liner and a nitrile micro-foam coating, these gloves offer high dexterity and enhanced grip while providing ANSI/ISEA A3 cut protection. The lightweight, fiberglass-free design and breathable construction ensure comfort during all-day wear, and the gloves are machine washable for extended service life.
  • Engineered with CoreShield™ yarn and a durable nitrile micro-foam coating
  • Touchscreen compatible and ESD-safe for use with electronic devices
  • Reinforced thumb crotch for added durability
  • Silicone-free and certified for food contact
  • Machine washable for cost-effective reusability
  • Color-coded for easy identification of cut protection levels
Catalog No. 50-005-08146