Learn More
MSC G-Tek CoreShield Seamless Knit Glove, HPPE, A3 Cut, Black, 100/Case
Supplier: MSC 37102472
These 15-gauge, fiberglass-free engineered yarn gloves provide reliable A3 cut resistance and comfort for a wide range of industrial applications. Featuring a breathable foam nitrile coating on the palms and fingers, these gloves are designed to disperse fluids for enhanced grip in dry and slightly oily environments. The seamless, touchscreen-compatible construction offers dexterity and is safe for food handling.
- Fiberglass-free and silicone-free design
- ANSI cut level A3 and EN 388 442C rating
- Breathable foam nitrile coating for superior dry and oily grip
- Touchscreen compatible and ESD-safe construction
- Food certified under FDA 21 CFR and EC No. 1935/2004
- Oeko-Tex® standard 100 certified
- Vend-ready packaging