Learn More
MSC PIP Cut Resistant Gloves, Size 8, A3, Foam Nitrile, 100/Case
Supplier:  MSC 37104387
Encompass
These size 8 cut-resistant gloves feature an 18-gauge liner constructed from recycled polyester, HPPE, carbon fiber, spandex, and nitrile foam. Designed for high-performance handling, they provide reliable protection in mechanical, assembly, and metal processing environments.
  • Cut resistance level A3 and EN 388 rating 4X42C
  • Foam nitrile coating on palm and fingers for superior grip
  • ESD safe construction compliant with EN 16350
  • Touchscreen compatible design
  • Certified for food contact under FDA 21 CFR 177.2600 and EC 1935/2004
  • Fiberglass-free and silicone-free material composition
  • Oeko-Tex® standard 100 certified
  • Reusable and launderable design
Catalog No. 50-005-08110