Learn More
MSC PIP Cut-Resistant Gloves, CORESHIELD 3.0, A2, 15 Gauge, Medium, 10/Pk.
Supplier:  MSC 37096013
Encompass
These cut-resistant gloves provide reliable protection and comfort for various industrial applications. Featuring a 15-gauge knit construction and a nitrile foam palm coating, they offer an enhanced grip in dry, wet, and oily conditions while maintaining dexterity. The seamless lining includes recycled polyester, carbon fiber, and performance PE fibers for a comfortable fit.
  • ANSI/ISEA cut resistance level A2 protection
  • Nitrile foam palm coating for improved dry, wet, and oil grip
  • Seamless lining for enhanced comfort and dexterity
  • Food contact certified according to EC 1935/2004
  • Anti-static properties for hazardous environments
  • Fiberglass-free construction
  • Elastic knit cuff for a secure fit
  • Overall length of 9.83 in.
Catalog No. 50-005-08104