Learn More
Cytek Biosciences Inc PE-Cy7 aMu CD62L Anti Primary
Supplier: Cytek Biosciences Inc 600621U100
Product Name: PE-Cyanine7 Anti-Mouse CD62L (L-Selectin) (MEL-14)
Product Class: Antibody
Type: Primary
Product Name: PE-Cyanine7 Anti-Mouse CD62L (L-Selectin) (MEL-14)
Cytek Catalog Number: 60-0621-U100
Marker: CD62L (L-Selectin)
Target Species: Mouse
Clonality: Monoclonal
Clone: MEL-14
Host Species: Rat
Isotype: IgG2a, k
Conjugation: PE-Cyanine7
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.