Learn More
Biolegend PE/Cyanine7 anti-human CD137 (4-1BB)
Supplier:  Biolegend 309817
Encompass_Preferred
PE/Cyanine7 anti-human CD137 (4-1BB) [4B4-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-535