Learn More
Biolegend PE/Cyanine7 anti-human/mouse Granzyme B Recombinant
Supplier:  Biolegend 372214
Encompass_Preferred
PE/Cyanine7 anti-human/mouse Granzyme B Recombinant [QA16A02]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests
Catalog No. NC1680129