Learn More
Biolegend PE/Cyanine7 anti-mouse CD3ε
Supplier:  Biolegend 100320
Encompass_Preferred
PE/Cyanine7 anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-396