Learn More
Biolegend PE/Cyanine7 anti-human CD162
Supplier:  Biolegend 328816
Encompass_Preferred
PE/Cyanine7 anti-human CD162 [KPL-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-272-2327